missing translation for 'onlineSavingsMsg'
Learn More

DOT1L Rabbit anti-Human, Mouse, Rat, Clone: S01-7F6, Novus Biologicals™

Product Code. 18361646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361646 100 μg 100µL
18341867 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18361646 Supplier Novus Biologicals Supplier No. NBP315025100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DOT1L Monoclonal antibody specifically detects DOT1L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOT1L
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S01-7F6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4
Host Species Rabbit
Immunogen Recombinant protein of human DOT1L (Uniprot#: Q8TEK3)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84444
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.