missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Dot Blot Hybridisation Manifold System
Description
- High density acrylic
- Microfiltration manifold
- Allows unfractionated samples to be immobilised onto membrane
- Immediate screening with nucleic acid probes or antibodies
- Upper manifold - allows sample to be loaded into each dot/slot
- Stainless steel thumbscrews and precision-lapped mating surfaces
- Template to allow membrane to be cut down to size
- Vacuum Port, can be connected to water aspirator or vacuum pump
- Vacuum reservoir to serve as collection chamber for filtrate
- Requires vacuum pump or water aspirator equivalent to 600mm Hg (0.8 bar)
- DHM-48: Used for 5 x 12.8cm membrane - 48 sample throughput
- DHM-96: Used for 7.5 x 11.3cm membrane - 96 sample throughput
- Compatible with multichannel pipettes and standard 96-well microtitre plates
Specifications
Specifications
| Device Holds | 48 Samples |
| Type | Dot Blot Hybridization System |
| Includes | 48-well Dot Blot Hybridisation Manifold in 3 x 16 array,paired upper dot-forming modules for dots3mm in diameter, and lower vacuum chamber module |
| No. of Slots | 48 |
| Temperature (Metric) Operating | 65°C |
| Volume (Metric) Sample | 100 μL |
| Dimensions (L x W x H) | D x W x H: 7.5 x 18.5 x 7 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?