Learn More
Mettler Toledo™ Dosing Unit for Excellence and Compact Potentiometric Titrators
Product Code. 17839384 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
CAN
Unit Size:
Each
Product Code. Connector Types unitSize
17839384 CAN Each
1 options
Product Code. 17839384 Supplier Mettler Toledo™ Supplier No. 51109030
Accessory for Excellence and Compact Potentiometric Titrators.
  • For extended dispensing and titration purposes
  • Plug & Play allows easy addition of Dosing Units to the titrator
  • Burettes are automatically recognized by the drive
  • Connection: CAN
  • Resolution (drive): 20'000 steps
TRUSTED_SUSTAINABILITY
Specifications
Connector Types CAN
Type Dosing Unit
For Use With (Application) Extended dispensing and titration purposes
Resolution 20'000 steps