missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSILAB

Product Code. 12860933
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
30 mL
60 mL
100 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12860933 2.5 mL Each
12870933 5 mL Each
12890933 10 mL Each
12880933 30 mL Each
12800943 60 mL Each
12810943 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12860933 Supplier KARTELL Supplier No. 8308200
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dosilab is a new series of adjustable volume bottle top dispenser for dosing chemicals with precision, accuracy and inexpensiveness. The six models of the new line are simple to use and easy to disassemble to perform common cleaning and maintenance operations.

  • Completely autoclavable at +121°C without need for disassembly.
  • PTFE piston with silicone O-ring ensuring a high degree of accuracy.
  • Internal cylinder in borosilicate glass protected by PP sheath.
  • Valve housing: PTFE body in a single piece ensuring high chemical resistance.
  • Accuracy and Reproducibility: according to the ISO 8655 standards.
  • Compatibility: excellent with all reagents including organic solvents and concentrated acids, except hydrofluoric acid (HF).
  • Recalibration: simple and quick with its special accessory.
  • All the internal components that during use are in contact with liquid solutions are made of PTFE, FEP and borosilicate glass. They therefore possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.

Specifications

Accuracy ± 0.015 mL
Angle 0⪚
Product Type Adjustable Volume Dispenser
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Color White
Channels 1
Material FEP, PTFE, Borosilicate Glass, Polypropylene
Volume (Metric) 2.5 mL
Display Analog
For Use With (Equipment) Reagent Bottle
Includes 1 Bottle Top Dispenser
Increments 0.05 mL
Volume (English) 0.08 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.