missing translation for 'onlineSavingsMsg'
Learn More

Dorsomorphin dihydrochloride, MedChemExpress

Product Code. 16495668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16495668 10 mg 10mg
16461837 50 mg 50mg
16448367 5 mg 5mg
16406186 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16495668

Brand: Medchem Express HY13418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dorsomorphin dihydrochloride (BML-275 dihydrochloride; Compound C dihydrochloride) is a potent, selective and ATP-competitive AMPK inhibitor, with a Ki of 109 nM. Dorsomorphin dihydrochloride inhibits BMP pathway by targeting the type I receptors ALK2, ALK3, and ALK6. Dorsomorphin dihydrochloride induces autophagy.

Dorsomorphin dihydrochloride (BML-275 dihydrochloride

Specifications

Chemical Name or Material Dorsomorphin dihydrochloride
CAS 1219168-18-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27Cl2N5O
Quantity 10 mg
Synonym Compound C dihydrochlorideBML-275 dihydrochloride
Solubility Information H2O : 50 mg/mL (105.84 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (10.58 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.C12=C(C3=CC=NC=C3)C=NN1C=C(C4=CC=C(OCCN5CCCCC5)C=C4)C=N2
Molecular Weight (g/mol) 472.41
Formula Weight 472.41
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.