missing translation for 'onlineSavingsMsg'
Learn More

Doripenem monohydrate, MedChemExpress

Product Code. 16418398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16418398

Brand: Medchem Express HYB0187A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doripenem monohydrate is a new member of the carbapenem class of beta-lactam antibiotics with broad-spectrum coverage of Gram-positive, Gram-negative and anaerobic pathogens.

Doripenem monohydrate is a new member of the carbapenem class of beta-lactam antibiotics with broad-spectrum coverage of Gram-positive, Gram-negative and anaerobic pathogens.

Specifications

Chemical Name or Material Doripenem monohydrate
Physical Form Solid
CAS 364622-82-2
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H26N4O7S2
For Use With (Application) COVID-19-immunoregulation
Synonym S 4661 monohydrate
Solubility Information DMSO : 50 mg/mL (114.02 mM; Need ultrasonic) ∣H2O : 10 mg/mL (22.80 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(S[C@@H]3CN[C@H](CNS(=O)(N)=O)C3)[C@H](C)[C@]2([H])[C@@H]([C@H](O)C)C1=O)O.O
Molecular Weight (g/mol) 438.52
Formula Weight 438.52
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.