missing translation for 'onlineSavingsMsg'
Learn More

Doravirine, MedChemExpress

Product Code. 16473207
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473207

Brand: Medchem Express HY16767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doravirine (MK-1439) is a highly specific HIV-1 nonnucleoside reverse transcriptase inhibitor with IC50s of 4.5 nM, 5.5 nM and 6.1 nM against the wild type and K103N and Y181C reverse transcriptase mutants, respectively.

Doravirine (MK-1439) is a highly specific HIV-1 nonnucleoside reverse transcriptase inhibitor with IC50s of 4.5 nM, 5.5 nM and 6.1 nM against the wild type and K103N and Y181C reverse transcriptase mutants, respectively.

Specifications

Chemical Name or Material Doravirine
CAS 1338225-97-0
Color White
Purity Grade Notes Research
Molecular Formula C17H11ClF3N5O3
Quantity 50 mg
Synonym MK-1439
Solubility Information DMSO : 100 mg/mL (234.88 mM; Need ultrasonic)
SMILES N#CC1=CC(OC2=C(C(F)(F)F)C=CN(CC(N3C)=NNC3=O)C2=O)=CC(Cl)=C1
Molecular Weight (g/mol) 425.75
Formula Weight 425.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.