missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Doramectin

Product Code. 16574494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16574494 5 g 5g
16564494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16574494 Supplier Thermo Scientific Chemicals Supplier No. 465620050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117704-25-3
Molecular Formula C50H74O14
Molecular Weight (g/mol) 899.13
InChI Key QLFZZSKTJWDQOS-NJPOTTGESA-N
IUPAC Name (1'R,2S,4'S,5S,6R,8'R,10'E,13'S,14'E,16'E,20'R,21'R,24'S)-6-cyclohexyl-21',24'-dihydroxy-12'-{[(2R,4S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one
SMILES CO[C@H]1C[C@H](OC2[C@H](C)O[C@H](C[C@@H]2OC)OC2[C@@H](C)\C=C\C=C3/CO[C@@H]4[C@H](O)C(C)=C[C@@H](C(=O)O[C@H]5C[C@@H](C\C=C2/C)O[C@@]2(C5)O[C@H](C5CCCCC5)[C@@H](C)C=C2)[C@]34O)O[C@@H](C)[C@@H]1O

Specifications

Melting Point 116.0°C to 119.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: practically insoluble.
Formula Weight 899.11
Chemical Name or Material Doramectin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.