missing translation for 'onlineSavingsMsg'
Learn More

Doramectin, Mikromol™

Product Code. 18090530
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090530 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090530 Supplier Mikromol Supplier No. MM3612.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doramectin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doramectin
CAS 117704-25-3
Analyte or Component Names Doramectin
Shipping Condition Room Temperature
Molecular Formula C50 H74 O14
Certifications/Compliance ISO 17034
Synonym 25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)-avermectin A1a, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], avermectin A1a deriv., 25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin a1a, 25-Cyclohexylavermectin B1, Dectomax, Doramectin, Doramectin A1a, L 701023, UK 67994
InChI Formula InChI=1S/C50H74O14/c1-27-13-12-16-34-26-57-47-42(51)30(4)21-37(50(34,47)54)48(53)60-36-22-35(63-49(25-36)20-19-29(3)45(64-49)33-14-10-9-11-15-33)18-17-28(2)44(27)61-41-24-39(56-8)46(32(6)59-41)62-40-23-38(55-7)43(52)31(5)58-40/h12-13,16-17,19-21,27,29,31-33,35-47,51-52,54H,9-11,14-15,18,22-26H2,1-8H3/b13-12+,28-17+,34-16+/t27-,29-,31-,32-,35+,36-,37-,38-,39-,40-,41-,42+,43-,44-,45-,46-,47+,49+,50+/m0/s1
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]3[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O[C@H]6C[C@@H](C\C=C\3/C)O[C@@]7(C6)O[C@H](C8CCCCC8)[C@@H](C)C=C7)[C@]45O)C)O[C@@H](C)[C@@H]1O
Molecular Weight (g/mol) 899.11
Quantity 100 mg
Formula Weight 898.5079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.