missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doramectin, MedChemExpress
Description
Doramectin is a derivative of Ivermectin (HY-15310). Doramectin is a potent antiparasitic antibiotic. Doramectin is an active compound against S.mansoni in an NMRI mouse infection model.
Specifications
Specifications
| Chemical Name or Material | Doramectin |
| Physical Form | Solid |
| CAS | 117704-25-3 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C50H74O14 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 50 mg/mL (55.61 mM) |
| SMILES | CO[C@@H]1[C@@H](O[C@]2([H])O[C@@H](C)[C@H](O)[C@@H](OC)C2)[C@H](C)O[C@@]([H])(O[C@H](/C(C)=C/C[C@]3([H])C[C@]([H])(OC4=O)C[C@]5(C=C[C@H](C)[C@@H](C6CCCCC6)O5)O3)[C@@H](C)/C=C/C=C7CO[C@@]8([H])[C@]\7(O)[C@@]4([H])C=C(C)[C@H]8O)C1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?