missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: GIKHRIKWNR KALPSTAQIT EAQVAENRPG AFIKQGRKLD IDFGAEGNRY YEANYWQF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
This gene is found on chromosome 20, approximately 20 kbp downstream of the gene encoding cellular prion protein, to which it is biochemically and structurally similar. The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that is found predominantly in testis. Mutations in this gene may lead to neurological disorders.
Specifications
Specifications
| Antigen | Doppel |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Prnd |
| Gene Accession No. | Q9UKY0 |
| Gene Alias | AI450264; dJ1068H6.4; DOPPEL; doppelganger; DPL; prion gene complex, downstream; Prion protein 2; prion protein 2 (dublet); prion protein dublet; prion protein-like protein; prion-like protein doppel; PRND; PrPLP; UNQ1830/PRO3443 |
| Gene Symbols | Prnd |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
