missing translation for 'onlineSavingsMsg'
Learn More

Doppel Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18403039 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403039 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18403039 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP297004JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Doppel Polyclonal antibody specifically detects Doppel in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Doppel
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias dJ1068H6.4, DOPPEL, DPLprion gene complex, downstream, MGC41841, Prion protein 2, prion protein 2 (dublet), PrPLPprion-like protein doppel
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Doppel (Accession#: NP_036541.2; Met1-Gly152)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23627
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.