missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOPEY1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25569-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the dopey family. It is a transporter protein, possibly involved in protein traffic between late Golgi and early endosomes. This gene plays a potential role in functional brain alterations and in the pathogenesis of mental retardation in Down syndrome, it overexpression in the brain regions, that are altered in Down syndrome patients and involved in learning and memory processes.Specifications
| DOPEY1 | |
| Polyclonal | |
| Unconjugated | |
| DOPEY1 | |
| dopey family member 1, DOPEY1, KIAA1117, Protein dopey 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23033 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JWR5 | |
| DOPEY1 | |
| DOPEY1 Fusion Protein Ag22244 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction