missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dopamine Transporter Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14656
This item is not returnable.
View return policy
Description
This antibody is specific for the ~88 kDa DAT protein in Western blots of SDS solubilized human and mouse striatal samples and in IHC applications with formaldehyde-fixed human and monkey (Macaque) brain sections.
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications
| Dopamine Transporter | |
| Polyclonal | |
| Unconjugated | |
| Slc6a3 | |
| DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13162, 24898, 6531 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.30 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P23977, Q01959, Q61327 | |
| Slc6a3 | |
| Synthetic peptide corresponding to amino acid residues from the intracellular C-terminal region of human SLC6A3 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction