missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dopamine Transporter Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106751
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC6A3.
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications
| Dopamine Transporter | |
| Polyclonal | |
| Unconjugated | |
| Slc6a3 | |
| DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13162, 24898, 6531 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23977, Q01959, Q61327 | |
| Slc6a3 | |
| A synthesized peptide derived from human SLC6A3(Accession Q01959), corresponding to amino acid residues G481-F531. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction