missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine Receptor D4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92717-0.1ml
This item is not returnable.
View return policy
Description
Dopamine Receptor D4 Polyclonal antibody specifically detects Dopamine Receptor D4 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Dopamine Receptor D4 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| D(2C) dopamine receptor, D(4) dopamine receptor, D4DR, Dopamine D4 receptor, dopamine receptor D4, seven transmembrane helix receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human DRD4 (NP_000788.2). AWLLSPRLCDALMAMDVMLCTASIFNLCAISVDRFVAVAVPLRYNRQGGSRRQLLLIGATWLLSAAVAAPVLCGLNDVRGRDPAVCRLEDRDYVVYSSVCS | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 1815 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction