missing translation for 'onlineSavingsMsg'
Learn More

Dopamine Hydrochloride, TRC

Product Code. 30007678
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007678 1 g 1g
30008432 25 g 25g
18032130 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007678 Supplier Toronto Research Chemicals Supplier No. TRCD5337801G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8 H11 N O2 . Cl H
Molecular Weight (g/mol) 189.64
Synonym Dopamine Hydrochloride, Noradrenaline Tartrate Imp. C (EP) as Hydrochloride, Dobutamine Hydrochloride Imp. A (EP) as Hydrochloride, 4-(2-Aminoethyl)pyrocatechol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxy-β-phenethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride, ASL 279, Cardiosteril, Dopadic, Dopamine Fresenius, Dopamine Pierre Faba, Dopamine chloride, Dopamine hydrochloride, Dopastat, Dopmin, Dynatra, Inotropin, Inovan, Intropin, KW 3160, P 498, Rascordin, Revivan, Tensamin, m-Hydroxytyramine hydrochloride, β-(3,4-Dihydroxyphenyl)ethylamine hydrochloride
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES Cl.NCCc1ccc(O)c(O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H
Formula Weight 189.0557
Percent Purity >95%
Chemical Name or Material Dopamine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.