missing translation for 'onlineSavingsMsg'
Learn More

Dopamine Hydrochloride, LoGiCal

Product Code. 30458667
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30458667 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30458667

Brand: LoGiCal LGCFOR0384.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dopamine hydrochloride
CAS 62-31-7
Analyte or Component Names Dopamine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H11 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Dobutamine Hydrochloride Imp. A (EP), Dobutamine Imp. A (EP), Noradrenaline Tartrate Imp. C (EP), Noradrenaline Imp. C (EP), Dopamine Hydrochloride, Noradrenaline Tartrate Imp. C (EP) as Hydrochloride, Dobutamine Hydrochloride Imp. A (EP) as Hydrochloride, 4-(2-Aminoethyl)pyrocatechol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxy-beta-phenethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride, ASL 279, Cardiosteril, Dopadic, Dopamine Fresenius, Dopamine Pierre Faba, Dopamine chloride, Dopamine hydrochloride, Dopastat, Dopmin, Dynatra, Inotropin, Inovan, Intropin, KW 3160, P 498, Rascordin, Revivan, Tensamin, m-Hydroxytyramine hydrochloride, beta-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, Dobutamine Hydrochloride Impurity A as Hydrochloride, Noradrenaline Tartrate Impurity C as Hydrochloride, Dobutamine Impurity A as Hydrochloride, Noradrenaline Impurity C as Hydrochloride
InChI Formula InChI=1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H
Solution Type Neat
SMILES Cl.NCCc1ccc(O)c(O)c1
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 189.64
Quantity 10 mg
Formula Weight 189.0557 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.