missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D5R/DRD5 Antibody (SG4-D1b), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60019H
This item is not returnable.
View return policy
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Dopamine D5R/DRD5 | |
| Monoclonal | |
| HRP | |
| D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat. Reactivity with mouse has had mixed results. It does not react with human protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry | |
| SG4-D1b | |
| Western Blot, Immunohistochemistry | |
| DRD5 | |
| Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-Prot# P25115] | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 1816 | |
| Mouse, Rat, Human (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction