missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D3R/DRD3 Antibody (SR1747), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £358.00
Specifications
| Antigen | Dopamine D3R/DRD3 |
|---|---|
| Clone | SR1747 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672065
|
Novus Biologicals
NBP3-21963-100ul |
100 μg |
£358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643315
|
Novus Biologicals
NBP3-21963-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dopamine D3R/DRD3 Monoclonal antibody specifically detects Dopamine D3R/DRD3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Dopamine D3R/DRD3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205 | |
| A synthesized peptide derived from human Dopamine D3R/DRD3 (Uniprot #: P35462) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1747 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1814 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title