missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D2R/DRD2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | Dopamine D2R/DRD2 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630671
|
Novus Biologicals
NBP2-92097-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679230
|
Novus Biologicals
NBP2-92097-0.1ml |
0.1 mL |
£368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dopamine D2R/DRD2 Polyclonal antibody specifically detects Dopamine D2R/DRD2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Dopamine D2R/DRD2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 1813 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, dopamine receptor D2 isoform, seven transmembrane helix receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Dopamine D2R/DRD2 (NP_000786.1). LERQNWSRPFNGSDGK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title