missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine D1R/DRD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£343.00 - £497.00
Specifications
| Antigen | Dopamine D1R/DRD1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18307455
|
Novus Biologicals
NBP3-17644-25UL |
25 μg |
£343.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18313076
|
Novus Biologicals
NBP3-17644-100UL |
100 μg |
£497.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dopamine D1R/DRD1 Polyclonal antibody specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Dopamine D1R/DRD1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Neurotransmission | |
| PBS, pH 7.2, 40% glycerol | |
| 1812 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FRKAFSTLLGCYRLCPATNNAIETVSINNNGAAMFSSHHEPRGSISKECNLVYLIPHAVGSSEDLKKEEAAGIARPLEKLSPALSVILDYDTDVSLEKIQPITQNGQHP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title