missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dopamine beta-Hydroxylase Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human DBH. The Dopamine beta-Hydroxylase Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 1621 |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Symbol | DBH |
| Molecular Weight (g/mol) | 31 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54189. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction