Learn More
Invitrogen™ Dopamine beta Hydroxylase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA118314
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. Store lyophilized antibody at 2-8 °C. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.
The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.
Specifications
| Dopamine beta Hydroxylase | |
| Polyclonal | |
| Unconjugated | |
| DBH | |
| Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Rat, Bovine, Guinea Pig | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Neutralization, Western Blot | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| P09172, P15101, Q05754 | |
| DBH | |
| Native enzyme prepared from bovine adrenal glands. | |
| RUO | |
| 100727873, 1621, 25699, 280758 | |
| 4°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.