missing translation for 'onlineSavingsMsg'
Learn More

Dopamine beta-Hydroxylase Antibody (DBH/7224) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-26921JF635

Product Code. 30168526

  • £509.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dopamine beta-Hydroxylase Monoclonal antibody specifically detects Dopamine beta-Hydroxylase in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dopamine beta-Hydroxylase
Monoclonal
Janelia Fluor 635
DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1
Recombinant fragment (around aa200-400) of human Dopamine beta-Hydroxylase protein (exact sequence is proprietary)
0.1 mL
Cancer, Lipid and Metabolism, Neuroscience
1621
Store at 4°C in the dark.
IgG2 κ
Immunohistochemistry (Paraffin)
DBH/7224
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.