missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOPA decarboxylase Recombinant Rabbit Monoclonal Antibody (1H21L22)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703686
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Rat.
DOPA Decarboxylase (aromatic L-amino acid decarboxylase, AADC; DDC) catalyzes the second reaction in the biosynthesis of catecholamines and serotonin (Waymire and Haycock, 2002; Berry et al., 1996; Haycock et al., 2003). It is also involved in the biosynthesis of trace amines. DDC antibodies can therefore be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999).
Specifications
| DOPA decarboxylase | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O88533, P20711 | |
| DDC | |
| Peptides corresponding to human DOPA decarboxylase [1) aa431-aa448 2) aa49-aa65]. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1H21L22 | |
| Unconjugated | |
| DDC | |
| AADC; aromatic L-amino acid decarboxylase; aromatic-L-amino acid decarboxylase; aromatic-L-amino-acid decarboxylase; DDC; DOPA decarboxylase; dopa decarboxylase (aromatic L-amino acid decarboxylase); HGNC:2719; L-Dopa decarboxylase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13195, 1644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction