missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopa Decarboxylase/DDC Rabbit anti-Human, Clone: 40, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89567-50UL
This item is not returnable.
View return policy
Description
Dopa Decarboxylase/DDC Monoclonal specifically detects Dopa Decarboxylase/DDC in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| Dopa Decarboxylase/DDC | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 40 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28 | |
| Recombinant Human Dopa Decarboxylase/DDC protein | |
| 50 μg | |
| Neuroscience | |
| 1644 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction