Learn More
BINDER™ Door Lock with Key
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65, KB 130, KB 260, KB 470
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 720/1060/1600
KBF 130, KBF 260, KBF 470
KBF 720, KBF 1060, KBF 1600
KBF PRO 1060, KBF PRO 1600
KBF PRO 260/470/720
Unit Size:
Each
Description
Door is locked with a key to ensure secure access to samples.
Specifications
Specifications
| For Use With (Equipment) | KB 720, KB 1060 |
| Material | Steel |
| Type | Locking Latch |