Learn More
BINDER™ Door Lock with Key
Product Code. 30238293 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65, KB 130, KB 260, KB 470
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 720/1060/1600
KBF 130, KBF 260, KBF 470
KBF 720, KBF 1060, KBF 1600
KBF PRO 1060, KBF PRO 1600
KBF PRO 260/470/720
Unit Size:
Each
Product Code. 30238293
Brand: BINDER™ 80122529
Only null left
Add to basket
Add to basket
Lockable door handle with key.
Door is locked with a key to ensure secure access to samples.
Specifications
For Use With (Equipment) KB 65, KB 130, KB 260, KB 470
Material Steel
Type Locking Latch