Learn More
BINDER™ Door Lock and Door Handle
Product Code. 17804471 Shop All BINDER Products
Click to view available options
Includes:
Lockable door handle
Unit Size:
Each
Product Code. 17804471
Brand: BINDER™ 80122237
Only null left
Add to basket
Add to basket
Door lock and lockable door handle for use with the Series FP Avantgarde.Line Drying and Heating Chambers.
  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
For Use With (Equipment) BINDER Series FP Avantgarde.Line Drying and Heating Chambers
Type Door Lock
Includes Lockable door handle