Learn More
BINDER™ Door Handle Lock
Product Code. 15622066 Shop All BINDER Products
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
Product Code. Material unitSize
15622066 Plastic Each
1 options
Product Code. 15622066 Supplier BINDER™ Supplier No. 80121025
For Use With: BINDER units BD 56, BD 115, BD 260
  • Provides security for incubators and chambers
  • Lockable door handle
Specifications
Description Door lock - lockable door handle
For Use With (Equipment) BINDER Chambers BD 56, BD 115, BD 260
Material Plastic
Type Door Handle Lock