missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dooku 1
GREENER_CHOICE

Product Code. 18759816
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759816

Brand: Tocris Bioscience™ 6568/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversibly blocks Yoda1 activity; does not affect constitutive Piezo1 activity

Dooku 1 reversibly blocks Yoda1 (Cat. No. 5586) activity (IC50 = ∼1.5 μM). Exhibits no effect on constitutive Piezo1 channel activity, but antagonizes Yoda1-induced Piezo1 activation in endothelial cells, and inhibits Yoda1-induced Ca2+-entry in vitro. Also blocks Yoda1-induced relaxation of aorta ex vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2,6-Dichlorobenzyl)thio)-5-(1 H-pyrrol-2-yl)-1,3,4-oxadiazole
CAS 2253744-54-4
Quantity 10 mg
Target Other Piezo Channel Compounds
Molecular Formula C13H9Cl2N3OS
Formula Weight 326.2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.