missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Donovan S3 Shoe

Product Code. 17532646
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17532646 39 1 pair
17542646 40 1 pair
17552646 41 1 pair
17562646 42 1 pair
17572646 43 1 pair
17582646 44 1 pair
17592646 45 1 pair
17502656 46 1 pair
17512656 47 1 pair
17522656 48 1 pair
10 options
This item is not returnable. View return policy

Product Code. 17532646

Brand: Emma Safety Footwear MM99456939

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Donovan.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Brown
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 39
Fit Extra-wide Width
Ankle Style High
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.