missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP2-60650
158.10 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free with 0.05% Sodium Azide | |
| Purified Rabbit IgG (H&L) | |
| 1 mg | |
| Secondary | |
| Reconstitute with sterilized deionized water. Centrifuge to remove any particulates. | |
| Store at 4C after rehydration. For extended storage after rehydration, add an equal volume of glycerol and store at -20C. Avoid repeated freeze-thaw cycles. |
| Flow Cytometry, Immunomicroscopy | |
| 4.5mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Donkey | |
| Affinity purified | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins | |
| Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction