missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Sheep IgG (H+L) Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 |
| Host Species | Donkey |
| Immunogen | Sheep IgG whole molecule |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?