Learn More
Donkey anti-Sheep IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Donkey Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75428
Product Code. 18597702
  • £266.00 / 2mg
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Biotin
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
This Donkey anti-Sheep IgG (H+L) Secondary Antibody [Biotin] was developed against purified sheep IgG (H&L).
2 mg
Adaptive Immunity, Immunology
Sheep
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.5 mg/mL
Western Blot 1:2000 - 1:20000, ELISA 1:2000 - 1:20000, Immunohistochemistry 1:500 - 1:5000, Immunohistochemistry-Paraffin
Donkey
Affinity purified
RUO
Secondary
Store at 4°C. Do not freeze.