missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, Alkaline Phosphatase (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-75378
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Donkey | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.50mg/mL | |
| Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin | |
| Purified Rat IgG (H&L) | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins. This antibody has been pre-adsorbed against bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit and sheep IgG. | |
| Store at 4C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20C. Avoid repeated freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction