Learn More
Invitrogen™ Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin
Donkey Polyclonal Secondary Antibody
Brand:  Invitrogen™ A18749
Product Code. 15275294
  • £211.00 / 2mg
Description
Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody for ELISA, Flow, ICC/IF, Western Blot
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L) Highly Cross-Adsorbed
Polyclonal
Biotin
Donkey
Affinity chromatography
RUO
Rat
Secondary Antibody
IgG
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
1.5 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.2
Gamma Immunoglobins Heavy and Light chains.
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.