Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790
Description
This secondary antibody is designed for fluorescent Western blot detection on various near-infrared fluorescence instruments. This antibody can be used for multi-color and multiplexing detection when using other antibodies conjugated to compatible Alexa Fluor™ dyes and wavelengths. Other applications of this antibody include immunofluorescent and fluorescent imaging applications when using instrumentation with appropriate excitation and detection capabilities. This antibody shows minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat, and sheep serum proteins. Product will be shipped at Room Temperature.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Highly Cross-Adsorbed |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor 790 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Donkey |
| Immunogen | Gamma Immunoglobin. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.