Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 625, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen Q22085
Code : 50
Additional Details : Weight : 0.01000kg
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Mouse IgG (H+L) | |
Polyclonal | |
Qdot 625 | |
Donkey | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 μM | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Gamma Immunoglobins Heavy and Light chains | |
100 μL | |
Secondary | |
4° C, store in dark | |
IgG |