missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB120-7061
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:20000, ELISA 1:20000 - 1:80000, Immunohistochemistry 1:1000 - 1:5000 | |
| Donkey anti-Mouse IgG (H+L) Secondary Antibody [HRP] (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in donkey. | |
| 0.2 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction