Learn More
Invitrogen™ Donkey anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen™ SA1100
Description
Product SA1-100 is conjugated to HRP and optimized for ECL applications. This antibody reacst with the heavy and light chains of the immunoglobulin molecule. Pre-absorbed with the following species proteins: Bovine, chicken, goat, guinea pig, syrian hamster, horse, human, rabbit, and sheep serum proteins.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| HRP | |
| Donkey | |
| 250 μL | |
| Secondary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 40 μg/mL | |
| PBS with 50% glycerol, proprietary stabilizer and 0.07% Kathon | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
Safety and Handling
- Donkey anti-Mouse IgG (H+L) Cross Adsorbed Secondary Antibody, HRPconjugate
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methylisothiazol-3(2H)-one and 2-methylisothiazol-3(2H)-one
Your input is important to us. Please complete this form to provide feedback related to the content on this product.