Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Texas Red (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand:  Novus Biologicals NB120-7123
237.15 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18709323
  • £297.00 / 0.50mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Texas Red
Donkey
Multi-step
RUO
Secondary
Goat
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Goat IgG whole molecule
0.5 mg
Adaptive Immunity, Immunology
This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit and Rat Serum Proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.