Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Donkey Polyclonal Antibody
Brand:  Novus Biologicals NBP2-60656JF549
Product Code. 18451666
  • £306.00 / 0.50mL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
50mM Sodium Borate
Donkey anti-Goat IgG (H+L) Secondary Antibody was produced by repeated immunization with goat IgG whole molecule in donkey
0.5 mL
Adaptive Immunity, Immunology
Goat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Dot Blot
Janelia Fluor 549
Donkey
Multi-step
RUO
Secondary
Store at 4°C in the dark.