missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, DyLight 550, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-75601
159.80 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifica
| IgG (H+L) | |
| Polyclonal | |
| DyLight 550 | |
| Donkey | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| Purified Goat IgG (H&L) | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on goat IgG and light chains on all goat immunoglobulins | |
| Goat | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto