missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NB120-7124
205.70 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:60000, ELISA 1:600000, Immunohistochemistry 1:1000 - 1:5000 | |
| Donkey anti-Goat IgG (H+L) Secondary Antibody [Biotin] (Pre-adsorbed) was produced by repeated immunization with goat IgG whole molecule in donkey. | |
| 0.2 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Horse, Hamster, Mouse, Rabbit and Rat Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Goat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction