missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Goat IgG (H+L) Secondary Antibody, AP
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen™ PA130370
This item is not returnable.
View return policy
Description
PA1-30370 detects Goat IgG from goat samples.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Goat IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Donkey | |
| IgG fraction | |
| RUO | |
| Goat | |
| Secondary Antibody | |
| IgG |
| Dot Blot, ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.001M MgCl2, 0.0001M ZnCl2, 50% glycerol, 1% BSA and 0.09% sodium azide; pH 8 | |
| Goat IgG whole molecule. | |
| 1 mg | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction