missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donepezil, MedChemExpress
Description
Donepezil (E2020 free base) is a specific and potent AChE inhibitor with IC50s of 8.12 nM and 11.6 nM for bovine AChE and human AChE, respectively.
Specifications
Specifications
| Chemical Name or Material | Donepezil |
| CAS | 120014-06-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H29NO3 |
| Quantity | 10 mg |
| Synonym | E2020 free base |
| Solubility Information | DMSO : 33.33 mg/mL (87.83 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.27 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(C(C=C(OC)C(OC)=C1)=C1C2)C2CC(CC3)CCN3CC4=CC=CC=C4 |
| Molecular Weight (g/mol) | 379.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?