missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donepezil Hydrochloride, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Description
Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE. Donepezil exhibits neuroprotective effect on Aβ42 neurotoxicity.
Specifications
Specifications
| Chemical Name or Material | Donepezil Hydrochloride |
| CAS | 120011-70-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30ClNO3 |
| Quantity | 100 mg |
| Synonym | E2020 |
| Solubility Information | H2O : 33.33 mg/mL (80.13 mM; Need ultrasonic) ∣DMSO : 6.2 mg/mL (14.91 mM; Need warming) |
| SMILES | O=C1C(CC2CCN(CC3=CC=CC=C3)CC2)CC4=C1C=C(OC)C(OC)=C4.[H]Cl |
| Molecular Weight (g/mol) | 415.95 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction