missing translation for 'onlineSavingsMsg'
Learn More

Donepezil Hydrochloride, MedChemExpress

Product Code. 16454217
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454217 50 mg 50mg
16418076 100 mg 100mg
16411366 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16454217 Supplier Medchem Express Supplier No. HYB0034
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE. Donepezil exhibits neuroprotective effect on Aβ42 neurotoxicity.

Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE. Donepezil exhibits neuroprotective effect on Aβ42 neurotoxicity.

Specifications

Chemical Name or Material Donepezil Hydrochloride
CAS 120011-70-3
Color White
Purity Grade Notes Research
Molecular Formula C24H30ClNO3
Quantity 50 mg
Synonym E2020
Solubility Information H2O : 33.33 mg/mL (80.13 mM; Need ultrasonic) ∣DMSO : 6.2 mg/mL (14.91 mM; Need warming)
SMILES O=C1C(CC2CCN(CC3=CC=CC=C3)CC2)CC4=C1C=C(OC)C(OC)=C4.[H]Cl
Molecular Weight (g/mol) 415.95
Formula Weight 415.95
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.